SY3H3D681C69 Synthonix Corporation

2-(Pyridin-3-yl)pyridin-4-amine

  • MDL number No MDL found
  • Cas Number : 40963-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53872.html

Safety Information

MSDS https://synthonix.com/53872.html
Availability details for
Quantity
  • Availability
  • Add to Cart