SY3H3D681C6A Synthonix Corporation

Phenyl (3-methoxy-1,2,4-thiadiazol-5-yl)carbamate

  • MDL number MFCD30489554
  • Cas Number : 1445988-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53885.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53885.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart