SY3H3D681C6B Synthonix Corporation

N-PhenylsucciniMide

  • MDL number No MDL found
  • Cas Number : 83-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53916.html

Safety Information

MSDS https://synthonix.com/53916.html
Availability details for
Quantity
  • Availability
  • Add to Cart