SY3H3D681C71 Synthonix Corporation

(Perfluoro-1,4-phenylene)dimethanamine acetate

  • MDL number No MDL found
  • Cas Number : 1841081-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/54079.html

Safety Information

MSDS https://synthonix.com/54079.html
Availability details for
Quantity
  • Availability
  • Add to Cart