SY3H3D681C72 Synthonix Corporation

(4-(1H-Pyrazol-1-yl)phenyl)methanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1107632-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54103.html

Safety Information

MSDS https://synthonix.com/54103.html
Availability details for
Quantity
  • Availability
  • Add to Cart