SY3H3D681C7D Synthonix Corporation

1-(Pyridin-2-ylsulfonyl)piperidin-4-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1353958-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/54193.html

Safety Information

MSDS https://synthonix.com/54193.html
Availability details for
Quantity
  • Availability
  • Add to Cart