SY3H3D681C7E Synthonix Corporation

3-Isopropoxy-4-methylaniline hydrochloride

  • Cas Number : 1795507-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54195.html

Safety Information

MSDS https://synthonix.com/54195.html
Availability details for
Quantity
  • Availability
  • Add to Cart