SY3H3D681C7F Synthonix Corporation

(S)-3-(Piperidin-2-yl)pyridine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 31945-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54235.html

Safety Information

MSDS https://synthonix.com/54235.html
Availability details for
Quantity
  • Availability
  • Add to Cart