SY3H3D681C86 Synthonix Corporation

4-(Pyridin-4-yl)aniline

  • Cas Number : 13296-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54259.html

Safety Information

MSDS https://synthonix.com/54259.html
Availability details for
Quantity
  • Availability
  • Add to Cart