SY3H3D681C93 Synthonix Corporation

(2-(1H-Pyrrol-1-yl)phenyl)methanol

  • Cas Number : 61034-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54407.html

Safety Information

MSDS https://synthonix.com/54407.html
Availability details for
Quantity
  • Availability
  • Add to Cart