SY3H3D681C94 Synthonix Corporation

5-Isopropylindoline-2,3-dione

  • MDL number MFCD02947184
  • Cas Number : 150560-58-0
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/54410.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54410.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart