SY3H3D681C95 Synthonix Corporation

4-Isopropoxy-3-(trifluoromethyl)benzoic acid

  • MDL number No MDL found
  • Cas Number : 213598-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54469.html

Safety Information

MSDS https://synthonix.com/54469.html
Availability details for
Quantity
  • Availability
  • Add to Cart