SY3H3D681C96 Synthonix Corporation

N-(1-Phenylethyl)prop-2-yn-1-amine

  • MDL number MFCD03502452
  • Cas Number : 56862-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54604.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54604.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart