SY3H3D681C98 Synthonix Corporation

(S)-1-(4-Phenyl-1H-imidazol-2-yl)ethanamine

  • MDL number No MDL found
  • Cas Number : 864825-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54618.html

Safety Information

MSDS https://synthonix.com/54618.html
Availability details for
Quantity
  • Availability
  • Add to Cart