SY3H3D681C99 Synthonix Corporation

5-(4-Isopropylphenyl)-1,3,4-thiadiazol-2-amine

  • MDL number No MDL found
  • Cas Number : 100987-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54624.html

Safety Information

MSDS https://synthonix.com/54624.html
Availability details for
Quantity
  • Availability
  • Add to Cart