SY3H3D681C9B Synthonix Corporation

4-Isopropoxy-2-methylbenzonitrile

  • MDL number No MDL found
  • Cas Number : 1369807-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54630.html

Safety Information

MSDS https://synthonix.com/54630.html
Availability details for
Quantity
  • Availability
  • Add to Cart