SY3H3D681CC7 Synthonix Corporation

4-Phenoxypiperidine

  • MDL number No MDL found
  • Cas Number : 3202-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54852.html

Safety Information

MSDS https://synthonix.com/54852.html
Availability details for
Quantity
  • Availability
  • Add to Cart