SY3H3D681CCB Synthonix Corporation

2-(Piperidin-4-yl)phenol

  • Cas Number : 161609-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54856.html

Safety Information

MSDS https://synthonix.com/54856.html
Availability details for
Quantity
  • Availability
  • Add to Cart