SY3H3D681CCD Synthonix Corporation

3-(Piperidin-1-yl)phenol

  • MDL number No MDL found
  • Cas Number : 27292-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54857.html

Safety Information

MSDS https://synthonix.com/54857.html
Availability details for
Quantity
  • Availability
  • Add to Cart