SY3H3D681CD9 Synthonix Corporation

(2-(Piperidin-1-ylsulfonyl)phenyl)boronic acid

  • MDL number No MDL found
  • Cas Number : 957034-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54947.html

Safety Information

MSDS https://synthonix.com/54947.html
Availability details for
Quantity
  • Availability
  • Add to Cart