SY3H3D681CDD Synthonix Corporation

4-(Pentyloxy)phenol

  • MDL number No MDL found
  • Cas Number : 18979-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54954.html

Safety Information

MSDS https://synthonix.com/54954.html
Availability details for
Quantity
  • Availability
  • Add to Cart