SY3H3D681CDF Synthonix Corporation

Pentane-1,5-diyl diacrylate

  • MDL number No MDL found
  • Cas Number : 36840-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54968.html

Safety Information

MSDS https://synthonix.com/54968.html
Availability details for
Quantity
  • Availability
  • Add to Cart