SY3H3D681CE1 Synthonix Corporation

2,3,4,5,6-Pentamethylphenylboronic Acid

  • Cas Number : 1028205-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55003.html

Safety Information

MSDS https://synthonix.com/55003.html
Availability details for
Quantity
  • Availability
  • Add to Cart