SY3H3D681D03 Synthonix Corporation

4-(Pyrimidin-5-yl)benzoic acid

  • Cas Number : 216959-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55230.html

Safety Information

MSDS https://synthonix.com/55230.html
Availability details for
Quantity
  • Availability
  • Add to Cart