SY3H3D681D20 Synthonix Corporation

5-Phenylpyridin-2-ol

  • Cas Number : 76053-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55345.html

Safety Information

MSDS https://synthonix.com/55345.html
Availability details for
Quantity
  • Availability
  • Add to Cart