SY3H3D681D27 Synthonix Corporation

4-(1H-Pyrrol-1-yl)benzaldehyde

  • MDL number No MDL found
  • Cas Number : 23351-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55351.html

Safety Information

MSDS https://synthonix.com/55351.html
Availability details for
Quantity
  • Availability
  • Add to Cart