SY3H3D681D2D Synthonix Corporation

4-(Piperazin-1-yl)furo[3,2-c]pyridine hydrochloride

  • MDL number No MDL found
  • Cas Number : 119924-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55456.html

Safety Information

MSDS https://synthonix.com/55456.html
Availability details for
Quantity
  • Availability
  • Add to Cart