SY3H3D681D30 Synthonix Corporation

cis-4-Phenylthio-L-proline hydrochloride

  • Cas Number : 105107-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55470.html

Safety Information

MSDS https://synthonix.com/55470.html
Availability details for
Quantity
  • Availability
  • Add to Cart