SY3H3D681D36 Synthonix Corporation

3-(Pyridin-2-yl)aniline dihydrochloride

  • MDL number No MDL found
  • Cas Number : 1170936-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55547.html

Safety Information

MSDS https://synthonix.com/55547.html
Availability details for
Quantity
  • Availability
  • Add to Cart