SY3H3D681D40 Synthonix Corporation

Phenyl(pyridin-4-yl)methanamine

  • MDL number No MDL found
  • Cas Number : 58088-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/55701.html

Safety Information

MSDS https://synthonix.com/55701.html
Availability details for
Quantity
  • Availability
  • Add to Cart