SY3H3D681D41 Synthonix Corporation

4-Phenoxybenzene-1,2-diamine

  • MDL number No MDL found
  • Cas Number : 13940-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/55703.html

Safety Information

MSDS https://synthonix.com/55703.html
Availability details for
Quantity
  • Availability
  • Add to Cart