SY3H3D681D44 Synthonix Corporation

(E)-3-(2-Propyl-6-(trifluoromethyl)pyridin-3-yl)acrylic acid

  • MDL number No MDL found
  • Cas Number : 1005174-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/55750.html

Safety Information

MSDS https://synthonix.com/55750.html
Availability details for
Quantity
  • Availability
  • Add to Cart