SY3H3D681D47 Synthonix Corporation

7-Isopropyl-1H-indole-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 383132-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55817.html

Safety Information

MSDS https://synthonix.com/55817.html
Availability details for
Quantity
  • Availability
  • Add to Cart