SY3H3D681D48 Synthonix Corporation

(R)-4-Phenyl-3-propionyloxazolidin-2-one

  • Cas Number : 160695-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55827.html

Safety Information

MSDS https://synthonix.com/55827.html
Availability details for
Quantity
  • Availability
  • Add to Cart