SY3H3D681D49 Synthonix Corporation

(S)-4-Phenyl-3-propionyloxazolidin-2-one

  • Cas Number : 184363-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55828.html

Safety Information

MSDS https://synthonix.com/55828.html
Availability details for
Quantity
  • Availability
  • Add to Cart