SY3H3D681D4F Synthonix Corporation

8-(Piperazin-1-yl)quinazoline

  • Cas Number : 221377-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55889.html

Safety Information

MSDS https://synthonix.com/55889.html
Availability details for
Quantity
  • Availability
  • Add to Cart