SY3H3D681D50 Synthonix Corporation

N-(1H-Pyrazolo[3,4-b]pyridin-3-yl)cyclopentanecarboxamide

  • MDL number No MDL found
  • Cas Number : 1789536-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55891.html

Safety Information

MSDS https://synthonix.com/55891.html
Availability details for
Quantity
  • Availability
  • Add to Cart