SY3H3D681D53 Synthonix Corporation

1,4-Phenylene dipropionate

  • MDL number No MDL found
  • Cas Number : 7402-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55938.html

Safety Information

MSDS https://synthonix.com/55938.html
Availability details for
Quantity
  • Availability
  • Add to Cart