SY3H3D681D57 Synthonix Corporation

2-(4-Isopropoxy-3-methoxyphenyl)acetonitrile

  • MDL number No MDL found
  • Cas Number : 861069-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55980.html

Safety Information

MSDS https://synthonix.com/55980.html
Availability details for
Quantity
  • Availability
  • Add to Cart