SY3H3D681D59 Synthonix Corporation

2-Pentyl-1H-benzo[d]imidazole

  • MDL number No MDL found
  • Cas Number : 5851-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56019.html

Safety Information

MSDS https://synthonix.com/56019.html
Availability details for
Quantity
  • Availability
  • Add to Cart