SY3H3D681D5A Synthonix Corporation

4-Phenethylpiperazin-2-one

  • MDL number No MDL found
  • Cas Number : 23099-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56021.html

Safety Information

MSDS https://synthonix.com/56021.html
Availability details for
Quantity
  • Availability
  • Add to Cart