SY3H3D681D5B Synthonix Corporation

1-Phenylpiperidine-3-carboxamide

  • MDL number No MDL found
  • Cas Number : 58971-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56023.html

Safety Information

MSDS https://synthonix.com/56023.html
Availability details for
Quantity
  • Availability
  • Add to Cart