SY3H3D681D5F Synthonix Corporation

3-(Piperidin-1-ylmethyl)phenol

  • MDL number MFCD00800250
  • Cas Number : 73279-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56136.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56136.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart