SY3H3D681D61 Synthonix Corporation

(1-Phenylpiperidin-4-yl)methanol

  • MDL number No MDL found
  • Cas Number : 697306-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56138.html

Safety Information

MSDS https://synthonix.com/56138.html
Availability details for
Quantity
  • Availability
  • Add to Cart