SY3H3D681D64 Synthonix Corporation

1-(Isopropylamino)-3-phenoxypropan-2-ol

  • MDL number No MDL found
  • Cas Number : 7695-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56236.html

Safety Information

MSDS https://synthonix.com/56236.html
Availability details for
Quantity
  • Availability
  • Add to Cart