SY3H3D681D6B Synthonix Corporation

1H-Pyrrolo[3,2-h]quinoline-2-carboxylic acid

  • Cas Number : 58457-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56418.html

Safety Information

MSDS https://synthonix.com/56418.html
Availability details for
Quantity
  • Availability
  • Add to Cart