SY3H3D681D70 Synthonix Corporation

6-Phenyl-5H-pyrrolo[3,2-d]pyrimidine-2,4-diol

  • MDL number No MDL found
  • Cas Number : 34771-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56494.html

Safety Information

MSDS https://synthonix.com/56494.html
Availability details for
Quantity
  • Availability
  • Add to Cart