SY3H3D681D7C Synthonix Corporation

3-(Pyridin-2-yl)benzoic acid

  • MDL number No MDL found

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56518.html

Safety Information

MSDS https://synthonix.com/56518.html
Availability details for
Quantity
  • Availability
  • Add to Cart