SY3H3D681D86 Synthonix Corporation

(4-(Pyridin-2-yl)phenyl)methanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1498333-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56570.html

Safety Information

MSDS https://synthonix.com/56570.html
Availability details for
Quantity
  • Availability
  • Add to Cart