SY3H3D681D87 Synthonix Corporation

3-(Piperidin-4-yl)benzo[d]oxazol-2(3H)-one hydrochloride

  • Cas Number : 162045-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56580.html

Safety Information

MSDS https://synthonix.com/56580.html
Availability details for
Quantity
  • Availability
  • Add to Cart